Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitro-D-phenylalanine hydrate 97%

Supplier: AMBEED, INC
Warning

A129971-100G A129971-25G
77258-424EA 43 USD InStock
77258-424 77258-426 77258-428 77258-430
4-Nitro-D-phenylalanine hydrate 97%
4-Nitro-D-phenylalanine hydrate
Precaución: for Research Use Only


Formula: C₉H₁₀N₂O₄·H₂O
MW: 228.202 g/mol
MDL Number: MFCD00150962
CAS Number: 444777-67-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty