Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bicyclo[2.2.1]hept-5-en-2-yl methacrylate 97%

Supplier: AMBEED, INC
Warning

A130432-1G A130432-250MG
77234-480EA 70 USD InStock
77234-480 77234-482
Bicyclo[2.2.1]hept-5-en-2-yl methacrylate 97%
Bicyclo[2.2.1]hept-5-en-2-yl methacrylate
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂
MDL Number: MFCD30478749
CAS Number: 3232-07-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty