Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-(Succinylbis(oxy))dibenzoic acid 97%

Supplier: AMBEED, INC
Warning

A130829-1G A130829-250MG A130829-5G
77246-530EA 20 USD InStock
77246-530 77246-532 77246-534
2,2'-(Succinylbis(oxy))dibenzoic acid 97%
2,2'-(Succinylbis(oxy))dibenzoic acid
Precaución: for Research Use Only


Formula: C₁₈H₁₄O₈
MDL Number: MFCD00133271
CAS Number: 578-19-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty