Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-bromo-4-methyl-5-nitrobenzoate 97%

Supplier: AMBEED, INC
Warning

A131769-1G A131769-250MG A131769-25G A131769-5G
77272-040EA 9 USD InStock
77272-040 77272-042 77272-044 77272-046
Methyl 3-bromo-4-methyl-5-nitrobenzoate 97%
Methyl 3-bromo-4-methyl-5-nitrobenzoate
Precaución: for Research Use Only


Formula: C₉H₈BrNO₄
MW: 274.07 g/mol
MDL Number: MFCD07367995
CAS Number: 223519-08-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty