Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-m-toluic acid 98%

Supplier: AMBEED, INC
Warning

A132017-100G A132017-10G A132017-1G A132017-250MG A132017-25G A132017-5G
77258-506EA 429 USD InStock
77258-506 77258-508 77258-510 77258-512 77258-514 77258-516
6-Methoxy-m-toluic acid 98%
6-Methoxy-m-toluic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MDL Number: MFCD00045849
CAS Number: 25045-36-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty