Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD9496 98+%

Supplier: AMBEED, INC
Warning

A133273-10MG A133273-1MG A133273-25MG A133273-50MG A133273-5MG
77234-592EA 55 USD InStock
77234-592 77234-594 77234-596 77234-598 77234-600
AZD9496 98+%
AZD9496
Precaución: for Research Use Only


Formula: C₂₅H₂₅F₃N₂O₂
MDL Number: MFCD28902195
CAS Number: 1639042-08-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty