Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-o-xylene 99%

Supplier: AMBEED, INC
Danger

A133701-1G A133701-25G A133701-5G
77285-410EA 28.84 USD InStock
77285-410 77285-412 77285-414 77285-416
4-Fluoro-o-xylene 99%
4-Fluoro-o-xylene
Precaución: for Research Use Only


Formula: C₈H₉F
MW: 124.16
Density: 1
MDL Number: MFCD00039217
CAS Number: 452-64-2
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty