Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diphenyl-1H-imidazole 98%

Supplier: AMBEED, INC
Warning

A134289-10G A134289-1G A134289-25G A134289-5G
77246-668EA 43 USD InStock
77246-668 77246-670 77246-672 77246-674 77246-676
2,4-Diphenyl-1H-imidazole 98%
2,4-Diphenyl-1H-imidazole
Precaución: for Research Use Only


Formula: C₁₅H₁₂N₂
MDL Number: MFCD07772899
CAS Number: 670-83-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty