Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SKPin C1 98%

Supplier: AMBEED, INC
Warning

A134403-1MG A134403-5MG
77234-652EA 13 USD InStock
77234-652 77234-654
SKPin C1 98%
SKPin C1
Precaución: for Research Use Only


Formula: C₁₈H₁₃BrN₂O₄S₂
MW: 465.34 g/mol
MDL Number: MFCD03705291
CAS Number: 432001-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty