Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-2-methylacetic acid 97%

Supplier: AMBEED, INC
Warning

A134730-100MG A134730-1G A134730-250MG A134730-5G
77234-656EA 32 USD InStock
77234-656 77234-658 77234-660 77234-662
2-Cyano-2-methylacetic acid 97%
2-Cyano-2-methylacetic acid
Precaución: for Research Use Only


Formula: C₄H₅NO₂
MDL Number: MFCD14544816
CAS Number: 632-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty