Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-1-Benzyl-4-(ethoxycarbonyl)pyrrolidine-3-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A134754-100MG A134754-1G A134754-250MG
77246-688EA 17 USD InStock
77246-688 77246-690 77246-692
trans-1-Benzyl-4-(ethoxycarbonyl)pyrrolidine-3-carboxylic acid 97%
trans-1-Benzyl-4-(ethoxycarbonyl)pyrrolidine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₉NO₄
MDL Number: MFCD22689418
CAS Number: 895243-39-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty