Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5-Tribromo-2,4,6-trimethylbenzene 97%

Supplier: AMBEED, INC
Warning

A134872-10G A134872-1G A134872-250MG A134872-25G A134872-5G
77234-674EA 19 USD InStock
77234-674 77234-676 77234-678 77234-680 77234-682
1,3,5-Tribromo-2,4,6-trimethylbenzene 97%
1,3,5-Tribromo-2,4,6-trimethylbenzene
Precaución: for Research Use Only


Formula: C₉H₉Br₃
MDL Number: MFCD00155003
CAS Number: 608-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty