Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TRX818 Sodium salt 98+%

Supplier: AMBEED, INC
Warning

A135075-10MG A135075-1MG A135075-25MG A135075-5MG
77272-182EA 367.99 USD InStock
77272-182 77272-184 77272-186 77272-188
TRX818 Sodium salt 98+%
TRX818 Sodium salt
Precaución: for Research Use Only


Formula: C₁₆H₁₁FNNa₂O₆P
MDL Number: MFCD31725038
CAS Number: 1256037-62-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty