Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4aS,9bR)-Ethyl 6-bromo-5-(2-(methylamino)-2-oxoethyl)-3,4,4a,5-tetrahydro-1H-pyrido[4,3-b]indole-2(9bH)-carboxylate 97%

Supplier: AMBEED, INC
Warning

A135193-100MG A135193-1G A135193-250MG
77258-612EA 31 USD InStock
77258-612 77258-614 77258-616
(4aS,9bR)-Ethyl 6-bromo-5-(2-(methylamino)-2-oxoethyl)-3,4,4a,5-tetrahydro-1H-pyrido[4,3-b]indole-2(9bH)-carboxylate 97%
(4aS,9bR)-Ethyl 6-bromo-5-(2-(methylamino)-2-oxoethyl)-3,4,4a,5-tetrahydro-1H-pyrido[4,3-b]indole-2(9bH)-carboxylate
Precaución: for Research Use Only


Formula: C₁₇H₂₂BrN₃O₃
MDL Number: MFCD30730054
CAS Number: 2098497-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty