Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate 98,5%

Supplier: AMBEED, INC
Warning

A135692-100G A135692-25G A135692-500G
77246-718EA 6 USD InStock
77246-718 77246-720 77246-722
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate 98,5%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate
Precaución: for Research Use Only


Formula: C₁₆H₃₀O₄
MW: 286.41 g/mol
MDL Number: MFCD00059267
CAS Number: 6846-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty