Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GSK-2110183 ≥99%

Supplier: AMBEED, INC
Warning

A136023-10MG A136023-1G A136023-1MG A136023-2MG A136023-5MG
77234-714EA 94 USD InStock
77234-714 77234-716 77234-718 77234-720 77234-722
GSK-2110183 ≥99%
GSK-2110183
Precaución: for Research Use Only


Formula: C₁₈H₁₇Cl₂FN₄OS
MW: 427.32 g/mol
MDL Number: MFCD26961098
CAS Number: 1047644-62-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty