Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)-6-methylpyridine 97%

Supplier: AMBEED, INC
Danger

A136376-1G A136376-250MG A136376-5G
77234-742EA 35 USD InStock
77234-742 77234-744 77234-746
2-(Chloromethyl)-6-methylpyridine 97%
2-(Chloromethyl)-6-methylpyridine
Precaución: for Research Use Only


Formula: C₇H₈ClN
MDL Number: MFCD09055262
CAS Number: 3099-29-4
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty