Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate 98%

Supplier: AMBEED, INC
Warning

A136544-100G A136544-10G A136544-1G A136544-25G A136544-500G A136544-5G
77272-262EA 48 USD InStock
77272-262 77272-264 77272-266 77272-268 77272-270 77272-272
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate 98%
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate
Precaución: for Research Use Only


Formula: C₁₇H₂₃NO₆
MDL Number: MFCD00065569
CAS Number: 13574-13-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty