Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-methyl-5-nitronicotinonitrile 95%

Supplier: AMBEED, INC
Warning

A137203-100MG A137203-1G A137203-250MG
77234-752EA 33 USD InStock
77234-752 77234-754 77234-756
2-Chloro-6-methyl-5-nitronicotinonitrile 95%
2-Chloro-6-methyl-5-nitronicotinonitrile
Precaución: for Research Use Only


Formula: C₇H₄ClN₃O₂
MDL Number: MFCD16877634
CAS Number: 26820-34-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty