Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LY-2584702 tosylate 98+%

Supplier: AMBEED, INC
Warning

A137621-10MG A137621-1MG A137621-50MG A137621-5MG
77234-764EA 16 USD InStock
77234-764 77234-766 77234-768 77234-770
LY-2584702 tosylate 98+%
LY-2584702 tosylate
Precaución: for Research Use Only


Formula: C₂₈H₂₇F₄N₇O₃S
MW: 617.62 g/mol
MDL Number: MFCD29054739
CAS Number: 1082949-68-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty