Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BLU-554 97%

Supplier: AMBEED, INC
Warning

A137706-10MG A137706-1MG A137706-5MG
77258-714EA 43 USD InStock
77258-714 77258-716 77258-718
BLU-554 97%
BLU-554
Precaución: for Research Use Only


Formula: C₂₄H₂₄Cl₂N₄O₄
MDL Number: MFCD30496704
CAS Number: 1707289-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty