Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Regorafenib monohydrate 98%

Supplier: AMBEED, INC
Warning

A137952-100MG A137952-10MG A137952-50MG
77234-778EA 8 USD InStock
77234-778 77234-780 77234-782
Regorafenib monohydrate 98%
Regorafenib monohydrate
Precaución: for Research Use Only


Formula: C₂₁H₁₇ClF₄N₄O₄
MDL Number: MFCD17170366
CAS Number: 1019206-88-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty