Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-2-pyrrolidone 97%

Supplier: AMBEED, INC
Danger

A138081-1000G A138081-100G A138081-25G A138081-500G
77234-784EA 40 USD InStock
77234-784 77234-786 77234-788 77234-790
1-Ethyl-2-pyrrolidone 97%
1-Ethyl-2-pyrrolidone
Precaución: for Research Use Only


Formula: C₆H₁₁NO
MDL Number: MFCD00003199
CAS Number: 2687-91-4
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty