Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-methylpyrimidine 97%

Supplier: AMBEED, INC
Warning

A138211-10G A138211-1G A138211-250MG A138211-25G A138211-5G
77285-616EA 38 USD InStock
77285-616 77285-618 77285-620 77285-622 77285-624
4-Chloro-6-methylpyrimidine 97%
4-Chloro-6-methylpyrimidine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂
MW: 128.559 g/mol
MDL Number: MFCD02322991
CAS Number: 3435-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty