Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3,5-dimethylbenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A138338-100G A138338-10G A138338-25G A138338-500G A138338-5G
77246-794EA 24 USD InStock
77246-794 77246-796 77246-798 77246-800 77246-802
4-Hydroxy-3,5-dimethylbenzaldehyde 98%
4-Hydroxy-3,5-dimethylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MDL Number: MFCD00006946
CAS Number: 2233-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty