Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Ethyl 3-(4-(benzyloxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoate 97%

Supplier: AMBEED, INC
Warning

A139016-1G A139016-5G
77272-388EA 23 USD InStock
77272-388 77272-390 77272-392
(S)-Ethyl 3-(4-(benzyloxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoate 97%
(S)-Ethyl 3-(4-(benzyloxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoate
Precaución: for Research Use Only


Formula: C₂₃H₂₉NO₅
MDL Number: MFCD26407317
CAS Number: 127132-32-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty