Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenyl-1,2-dihydro-3H-pyrazol-3-one hydrochloride 95%

Supplier: AMBEED, INC
Warning

A139411-100MG A139411-250MG
77246-846EA 18 USD InStock
77246-846 77246-848
2-Phenyl-1,2-dihydro-3H-pyrazol-3-one hydrochloride 95%
2-Phenyl-1,2-dihydro-3H-pyrazol-3-one hydrochloride
Precaución: for Research Use Only


Formula: C₉H₉ClN₂O
MDL Number: MFCD30489080
CAS Number: 2044772-89-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty