Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Aminophenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A139527-100G A139527-10G A139527-25G A139527-500G
77246-858EA 27 USD InStock
77246-858 77246-860 77246-862 77246-864
(4-Aminophenyl)acetic acid 98%
(4-Aminophenyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₄(NH₂))CH₂COOH
MW: 151.16 g/mol
Boiling Pt: 322
MDL Number: MFCD00007916
CAS Number: 1197-55-3
Merck Index: 13,00462
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty