Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Nitroquinolin-6-ol 97%

Supplier: AMBEED, INC
Warning

A139869-100MG A139869-1G A139869-250MG A139869-5G
77246-870EA 20 USD InStock
77246-870 77246-872 77246-874 77246-876
8-Nitroquinolin-6-ol 97%
8-Nitroquinolin-6-ol
Precaución: for Research Use Only


Formula: C₉H₆N₂O₃
MDL Number: MFCD18448969
CAS Number: 5437-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty