Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Veratramine 98%

Supplier: AMBEED, INC
Warning

A139966-100MG A139966-10MG A139966-1MG A139966-50MG A139966-5MG
77234-878EA 68 USD InStock
77234-878 77234-880 77234-882 77234-884 77234-886
Veratramine 98%
Veratramine
Precaución: for Research Use Only


Formula: C₂₇H₃₉NO₂
MDL Number: MFCD00468124
CAS Number: 60-70-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty