Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Cyanomethyl)benzeneboronic acid 96%

Supplier: AMBEED, INC
Warning

A139997-100MG A139997-1G A139997-250MG
77234-888EA 15 USD InStock
77234-888 77234-890 77234-892
3-(Cyanomethyl)benzeneboronic acid 96%
3-(Cyanomethyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₈H₈BNO₂
MDL Number: MFCD03788026
CAS Number: 220616-39-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty