Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methoxybenzoic acid 97%

Supplier: AMBEED, INC
Warning

A140438-100G A140438-10G A140438-1G A140438-250MG A140438-25G A140438-5G
77234-906EA 210.01 USD InStock
77234-906 77234-908 77234-910 77234-912 77234-914 77234-916
2-Chloro-5-methoxybenzoic acid 97%
2-Chloro-5-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₈H₇ClO₃
MDL Number: MFCD00466261
CAS Number: 6280-89-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty