Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Amino-3-bromo-2-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A140791-100G A140791-25G A140791-5G
77285-718EA 41 USD InStock
77285-718 77285-720 77285-722
6-Amino-3-bromo-2-methylpyridine 98%
6-Amino-3-bromo-2-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD00068230
CAS Number: 42753-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty