Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GLPG-1690 98+%

Supplier: AMBEED, INC
Warning

A141252-1MG A141252-25MG A141252-50MG A141252-5MG A141252-10MG
77258-802EA 13 USD InStock
77258-802 77258-804 77258-806 77258-808 77747-994
GLPG-1690 98+%
GLPG-1690
Precaución: for Research Use Only


Formula: C₃₀H₃₃FN₈O₂S
MDL Number: MFCD31544330
CAS Number: 1628260-79-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty