Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[4-(Bromomethyl)phenyl]benzoate 98%

Supplier: AMBEED, INC
Danger

A141268-100G A141268-25G A141268-5G
77272-476EA 55 USD InStock
77272-476 77272-478 77272-480
2-[4-(Bromomethyl)phenyl]benzoate 98%
2-[4-(Bromomethyl)phenyl]benzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₃BrO₂
MW: 305.1665 g/mol
MDL Number: MFCD06200816
CAS Number: 114772-38-2
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty