Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiophene-2,5-dicarbonitrile 97%

Supplier: AMBEED, INC
Warning

A141466-100MG A141466-10G A141466-1G A141466-250MG A141466-5G
77234-994EA 14 USD InStock
77234-994 77234-996 77234-998 77235-000 77235-002
Thiophene-2,5-dicarbonitrile 97%
Thiophene-2,5-dicarbonitrile
Precaución: for Research Use Only


Formula: C₆H₂N₂S
MDL Number: MFCD22987914
CAS Number: 18853-40-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty