Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxypropiononitrile 97%

Supplier: AMBEED, INC
Warning

A141843-100G A141843-25G A141843-500G
77272-492EA 19 USD InStock
77272-492 77272-494 77272-496
3-Hydroxypropiononitrile 97%
3-Hydroxypropiononitrile
Precaución: for Research Use Only


Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 228
Melting Pt: -46
MDL Number: MFCD00002826
CAS Number: 109-78-4
Merck Index: 13,03828
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty