Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-p-phenylenediamine 97%

Supplier: AMBEED, INC
Warning

A142003-100G A142003-10G A142003-1G A142003-25G A142003-5G
77258-834EA 121 USD InStock
77258-834 77258-836 77258-838 77258-840 77258-842
2-Chloro-p-phenylenediamine 97%
2-Chloro-p-phenylenediamine
Precaución: for Research Use Only


Formula: C₆H₇ClN₂
MDL Number: MFCD00128931
CAS Number: 615-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty