Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chlorochroman-4-one 97%

Supplier: AMBEED, INC
Warning

A142130-100G A142130-10G A142130-1G A142130-25G A142130-5G
77258-850EA 367 USD InStock
77258-850 77258-852 77258-854 77258-856 77258-858
6-Chlorochroman-4-one 97%
6-Chlorochroman-4-one
Precaución: for Research Use Only


Formula: C₉H₇ClO₂
MDL Number: MFCD00024070
CAS Number: 37674-72-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty