Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Bis(4-cyanatophenyl)propane 97%

Supplier: AMBEED, INC
Warning

A142183-100G A142183-25G A142183-500G
77246-992EA 55 USD InStock
77246-992 77246-994 77246-996 77246-998
2,2-Bis(4-cyanatophenyl)propane 97%
2,2-Bis(4-cyanatophenyl)propane
Precaución: for Research Use Only


Formula: C₁₇H₁₄N₂O₂
MDL Number: MFCD00037423
CAS Number: 1156-51-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty