Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-5-phenyl-1H-pyrazole 95%

Supplier: AMBEED, INC
Warning

A142236-100MG A142236-1G A142236-250MG A142236-5G
77272-522EA 11 USD InStock
77272-522 77272-524 77272-526 77272-528
3-Methyl-5-phenyl-1H-pyrazole 95%
3-Methyl-5-phenyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂
MDL Number: MFCD00524752
CAS Number: 3347-62-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty