Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Menadione sodium bisulphite BR

Supplier: AMBEED, INC
Danger

A142509-100MG A142509-25G A142509-100G
77258-884EA 5 USD InStock
77258-884 77592-692 77615-138
Menadione sodium bisulphite BR
Menadione sodium bisulphite
Precaución: for Research Use Only


Formula: C₁₁H₉NaO₅S
MW: 276.24 g/mol
MDL Number: MFCD00051456
CAS Number: 130-37-0
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty