Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BAW2881 ≥98%

Supplier: AMBEED, INC
Warning

A142514-10MG A142514-25MG A142514-2MG A142514-5MG
77258-886EA 116 USD InStock
77258-886 77258-888 77258-890 77258-892 77258-894
BAW2881 ≥98%
BAW2881
Precaución: for Research Use Only


Formula: C₂₂H₁₅F₃N₄O₂
MDL Number: MFCD30343850
CAS Number: 861875-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty