Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-(2-amino-3-(4-bromobenzoyl)phenyl)acetate 98%

Supplier: AMBEED, INC
Danger

A142956-10MG A142956-1MG A142956-50MG
77247-012EA 19 USD InStock
77247-012 77247-014 77247-016
Sodium 2-(2-amino-3-(4-bromobenzoyl)phenyl)acetate 98%
Sodium 2-(2-amino-3-(4-bromobenzoyl)phenyl)acetate
Precaución: for Research Use Only


Formula: C₁₅H₁₁BrNNaO₃
MDL Number: MFCD03701673
CAS Number: 91714-93-1
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty