Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PF-04554878 99%

Supplier: AMBEED, INC
Warning

A143423-1MG A143423-5MG A143423-100MG A143423-50MG
77235-096EA 11 USD InStock
77235-096 77235-098 77746-008 77746-009
PF-04554878 99%
PF-04554878
Precaución: for Research Use Only


Formula: C₂₀H₂₁F₃N₈O₃S
MW: 510.49 g/mol
MDL Number: MFCD25977806
CAS Number: 1073154-85-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty