Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Bis(trifluoromethyl)benzoic acid 95%

Supplier: AMBEED, INC
Warning

A143979-250MG A143979-5G
77258-938EA 5 USD InStock
77258-938 77258-940
2,4-Bis(trifluoromethyl)benzoic acid 95%
2,4-Bis(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₄F₆O₂
MW: 258.12
Boiling Pt: 260
MDL Number: MFCD00042496
CAS Number: 32890-87-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty