Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-4-nitrophenylacetate 99%

Supplier: AMBEED, INC
Warning

A144188-1G A144188-5G
77235-136EA 20 USD InStock
77235-136 77235-138
Sodium-4-nitrophenylacetate 99%
Sodium-4-nitrophenylacetate
Precaución: for Research Use Only


Formula: C₈H₇NO₄
MW: 203.13 g/mol
MDL Number: MFCD00070537
CAS Number: 7063-24-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty