Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-iodobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A144837-10G A144837-250MG A144837-25G A144837-5G
77272-686EA 36 USD InStock
77272-686 77272-688 77272-690 77272-692
3-Fluoro-4-iodobenzoic acid 97%
3-Fluoro-4-iodobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄FIO₂
MW: 266.0100 g/mol
MDL Number: MFCD07774250
CAS Number: 825-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty