Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-methoxypyrimidine 98%

Supplier: AMBEED, INC
Warning

A145201-100G A145201-10G A145201-1G A145201-250MG A145201-25G A145201-5G
77258-982EA 87 USD InStock
77258-982 77258-984 77258-986 77258-988 77258-990 77258-992
2-Chloro-4-methoxypyrimidine 98%
2-Chloro-4-methoxypyrimidine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
MDL Number: MFCD00194055
CAS Number: 22536-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty