Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-4-fluorophenol 97%

Supplier: AMBEED, INC
Warning

A146878-100MG A146878-1G A146878-250MG A146878-5G
77259-058EA 20 USD InStock
77259-058 77259-060 77259-062 77259-064
2-Ethyl-4-fluorophenol 97%
2-Ethyl-4-fluorophenol
Precaución: for Research Use Only


Formula: C₈H₉FO
MDL Number: MFCD00067856
CAS Number: 398-71-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty